1-(4-(Benzyloxy)phenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63433-1G 63433-500MG 63433-5G 63433-25G
77472-300EA 286.34 USD
77472-300 77484-928 77495-480 77504-808
1-(4-(Benzyloxy)phenyl)-2,2,2-trifluoroethanone ≥97%
1-(4-(Benzyloxy)phenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₅H₁₁F₃O₂
MW: 280.2418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07380843
CAS Number: 137614-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR