3-Fluoro-4-isopropoxy-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63398-1G 63398-500MG 63398-5G
77472-292EA 685.98 USD
77472-292 77484-920 77495-472
3-Fluoro-4-isopropoxy-5-methoxybenzaldehyde ≥97%
3-Fluoro-4-isopropoxy-5-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₃FO₃
MW: 212.2175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761332
CAS Number: 1400643-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR