(2-Ethoxy-3,5-bis(trifluoromethyl)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

63335-1G 63335-500MG 63335-5G
77472-288EA 918.76 USD
77472-288 77484-914 77495-468
(2-Ethoxy-3,5-bis(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(2-Ethoxy-3,5-bis(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₀F₆OS
Storage Temperature: Ambient
MDL Number: MFCD34761297
CAS Number: 2807456-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR