1-(5-Ethyl-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63182-1G 63182-500MG 63182-5G
77472-284EA 511.91 USD
77472-284 77484-910 77495-462
1-(5-Ethyl-2-fluorophenyl)ethanone ≥95%
1-(5-Ethyl-2-fluorophenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD22493251
CAS Number: 1551616-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR