2-Ethoxy-1-fluoro-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

63068-1G 63068-500MG 63068-5G 63068-25G
77472-280EA 76.22 USD
77472-280 77484-906 77495-454 77504-792
2-Ethoxy-1-fluoro-3-methoxybenzene ≥95%
2-Ethoxy-1-fluoro-3-methoxybenzene

Precaución: For research use only

Formula: C₉H₁₁FO₂
MW: 170.1808 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28363187
CAS Number: 909302-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR