5-Formyl-2,3-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63048-1G 63048-500MG 63048-5G
77472-276EA 1185.53 USD
77472-276 77484-902 77495-450
5-Formyl-2,3-dimethoxybenzoic acid ≥97%
5-Formyl-2,3-dimethoxybenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₀O₅
Storage Temperature: Ambient
MDL Number: MFCD24488828
CAS Number: 27203-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR