2-(5-(Benzyloxy)-2-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

63020-1G 63020-500MG 63020-5G 63020-25G
77472-272EA 415.09 USD
77472-272 77484-898 77495-446 77504-786
2-(5-(Benzyloxy)-2-fluorophenyl)-1,3-dioxolane ≥97%
2-(5-(Benzyloxy)-2-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₆H₁₅FO₃
MW: 274.2869 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761179
CAS Number: 2807468-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR