Methyl 3,5-difluoro-4-formylbenzoate ≥95%

Supplier: AOBChem USA

Warning

62937-1G 62937-500MG 62937-5G 62937-25G
77472-268EA 159.65 USD
77472-268 77484-894 77495-442 77504-782
Methyl 3,5-difluoro-4-formylbenzoate ≥95%
Methyl 3,5-difluoro-4-formylbenzoate

Precaución: For research use only

Formula: C₉H₆F₂O₃
MW: 200.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28525711
CAS Number: 1415124-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR