1-Chloro-2-iodo-5-isopropoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62903-1G 62903-500MG 62903-5G
77472-264EA 716.88 USD
77472-264 77484-890 77495-438
1-Chloro-2-iodo-5-isopropoxy-4-methylbenzene ≥97%
1-Chloro-2-iodo-5-isopropoxy-4-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₂ClIO
MW: 310.5591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761116
CAS Number: 2901065-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR