2-Bromo-5-chloro-3-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

62871-1G 62871-500MG 62871-5G
77472-260EA 486.16 USD
77472-260 77484-886 77495-434
2-Bromo-5-chloro-3-methylbenzoic acid ≥95%
2-Bromo-5-chloro-3-methylbenzoic acid

Precaución: For research use only

Formula: C₈H₆BrClO₂
MW: 249.4890 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761088
CAS Number: 2092682-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR