4-Bromo-1-iodo-2-(methoxymethoxy)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
62806-1G
62806-500MG
62806-5G
62806-25G
77472-256EA
324.45
USD
77472-256
77484-882
77495-430
77504-778
4-Bromo-1-iodo-2-(methoxymethoxy)benzene ≥97%
4-Bromo-1-iodo-2-(methoxymethoxy)benzene
Precaución: For research use only
Formula:
C₈H₈BrIO₂ MW: 342.9563 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34166923 CAS Number: 2245019-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...