1-Bromo-4-chloro-5-fluoro-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

62750-1G 62750-500MG 62750-5G
77472-252EA 526.33 USD
77472-252 77484-878 77495-426
1-Bromo-4-chloro-5-fluoro-2-isopropoxybenzene ≥97%
1-Bromo-4-chloro-5-fluoro-2-isopropoxybenzene

Precaución: For research use only

Formula: C₉H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34761024
CAS Number: 1906924-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR