2-Chloro-4-ethoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62742-1G 62742-500MG 62742-5G
77472-248EA 653.02 USD
77472-248 77484-874 77495-422
2-Chloro-4-ethoxy-5-methylbenzaldehyde ≥97%
2-Chloro-4-ethoxy-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁ClO₂
MW: 198.6461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761017
CAS Number: 2385879-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR