(3-Chloro-5-ethoxy-2-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62712-1G 62712-500MG 62712-5G 62712-25G
77472-244EA 415.09 USD
77472-244 77484-870 77495-418 77504-772
(3-Chloro-5-ethoxy-2-fluorophenyl)(methyl)sulfane ≥97%
(3-Chloro-5-ethoxy-2-fluorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀ClFOS
Storage Temperature: Ambient
MDL Number: MFCD34814216
CAS Number: 2706350-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR