(4-Carbamoyl-3-methoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

62592-1G 62592-500MG 62592-5G 62592-25G
77472-240EA 428.48 USD
77472-240 77484-866 77495-412 77504-766
(4-Carbamoyl-3-methoxyphenyl)boronic acid ≥95%
(4-Carbamoyl-3-methoxyphenyl)boronic acid

Precaución: For research use only

Formula: C₈H₁₀BNO₄
MW: 194.9803 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29496645
CAS Number: 1313617-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR