2-Fluoro-1-iodo-5-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62468-1G 62468-500MG 62468-5G 62468-25G
77472-236EA 111.24 USD
77472-236 77484-862 77495-408 77504-762
2-Fluoro-1-iodo-5-methoxy-3-methylbenzene ≥97%
2-Fluoro-1-iodo-5-methoxy-3-methylbenzene

Precaución: For research use only

Formula: C₈H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD34760848
CAS Number: 2383552-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR