2-(3-Ethoxy-2,4-difluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

62430-1G 62430-500MG 62430-5G 62430-25G
77472-232EA 415.09 USD
77472-232 77484-858 77495-404 77504-758
2-(3-Ethoxy-2,4-difluorophenyl)-1,3-dioxolane ≥97%
2-(3-Ethoxy-2,4-difluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₂F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34760837
CAS Number: 2901065-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR