1-(Methoxymethoxy)-2-methyl-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

62386-1G 62386-500MG 62386-5G 62386-25G
77472-228EA 415.09 USD
77472-228 77484-854 77495-400 77504-756
1-(Methoxymethoxy)-2-methyl-3-(trifluoromethyl)benzene ≥97%
1-(Methoxymethoxy)-2-methyl-3-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₁₀H₁₁F₃O₂
MW: 220.1883 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760819
CAS Number: 2901065-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR