3-Acetyl-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62319-1G 62319-500MG 62319-5G
77472-224EA 504.7 USD
77472-224 77484-850 77495-394
3-Acetyl-5-methylbenzaldehyde ≥97%
3-Acetyl-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD22544350
CAS Number: 1393534-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR