1-(3-Bromo-5-chloro-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

62269-1G 62269-500MG 62269-5G 62269-25G
77472-220EA 421.27 USD
77472-220 77484-846 77495-390 77504-748
1-(3-Bromo-5-chloro-2-fluorophenyl)ethanone ≥95%
1-(3-Bromo-5-chloro-2-fluorophenyl)ethanone

Precaución: For research use only

Formula: C₈H₅BrClFO
MW: 251.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32693996
CAS Number: 2092624-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR