2-(Benzyloxy)-5-chloro-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62171-1G 62171-500MG 62171-5G 62171-25G
77472-216EA 163.77 USD
77472-216 77484-842 77495-386 77504-744
2-(Benzyloxy)-5-chloro-3-fluorobenzaldehyde ≥97%
2-(Benzyloxy)-5-chloro-3-fluorobenzaldehyde

Precaución: For research use only

Formula: C₁₄H₁₀ClFO₂
MW: 264.6794 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760750
CAS Number: 2901102-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR