1-Bromo-2-methoxy-3-(methoxymethoxy)-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

62137-1G 62137-500MG 62137-5G 62137-25G
77472-212EA 140.08 USD
77472-212 77484-838 77495-382 77504-742
1-Bromo-2-methoxy-3-(methoxymethoxy)-5-methylbenzene ≥95%
1-Bromo-2-methoxy-3-(methoxymethoxy)-5-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO₃
MW: 261.1124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760733
CAS Number: 2901096-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR