1-Bromo-3-fluoro-2-isopropoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61912-1G 61912-500MG 61912-5G 61912-25G
77472-204EA 172.01 USD
77472-204 77484-830 77495-374 77504-736
1-Bromo-3-fluoro-2-isopropoxy-5-methylbenzene ≥97%
1-Bromo-3-fluoro-2-isopropoxy-5-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
MW: 247.1040 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760622
CAS Number: 1643457-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR