1-Iodo-2-(methoxymethoxy)-4,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

61843-1G 61843-500MG 61843-5G 61843-25G
77472-200EA 209.09 USD
77472-200 77484-826 77495-370 77504-732
1-Iodo-2-(methoxymethoxy)-4,5-dimethylbenzene ≥95%
1-Iodo-2-(methoxymethoxy)-4,5-dimethylbenzene

Precaución: For research use only

Formula: C₁₀H₁₃IO₂
MW: 292.1134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760599
CAS Number: 2807471-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR