1-(2,3-Difluoro-4-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

61746-1G 61746-500MG 61746-5G 61746-25G
77472-196EA 197.76 USD
77472-196 77484-822 77495-366 77504-728
1-(2,3-Difluoro-4-methoxyphenyl)ethanone ≥97%
1-(2,3-Difluoro-4-methoxyphenyl)ethanone

Precaución: For research use only

Formula: C₉H₈F₂O₂
MW: 186.1554 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26687730
CAS Number: 1537613-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR