3-(2-Chloro-4-fluorophenyl)-3-hydroxypropanoic acid ≥95%

Supplier: AOBChem USA

Warning

61578-1G 61578-500MG 61578-5G
77472-192EA 692.16 USD
77472-192 77484-818 77495-362
3-(2-Chloro-4-fluorophenyl)-3-hydroxypropanoic acid ≥95%
3-(2-Chloro-4-fluorophenyl)-3-hydroxypropanoic acid

Precaución: For research use only

Formula: C₉H₈ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD32118726
CAS Number: 1695650-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR