3,5-Dichloro-2-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61449-1G 61449-500MG 61449-5G
77472-188EA 515 USD
77472-188 77484-814 77495-358
3,5-Dichloro-2-methoxy-1,1'-biphenyl ≥97%
3,5-Dichloro-2-methoxy-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₃H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD34760347
CAS Number: 857590-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR