3-(Methoxymethyl)-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

61312-1G 61312-500MG 61312-5G
77472-184EA 893.01 USD
77472-184 77484-810 77495-354
3-(Methoxymethyl)-2-methylbenzoic acid ≥95%
3-(Methoxymethyl)-2-methylbenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₂O₃
MW: 180.2004 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34168011
CAS Number: 2648939-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR