3-Bromo-6-chloro-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61292-1G 61292-500MG 61292-5G 61292-25G
77472-180EA 226.6 USD
77472-180 77484-806 77495-350 77504-722
3-Bromo-6-chloro-2-hydroxybenzaldehyde ≥97%
3-Bromo-6-chloro-2-hydroxybenzaldehyde

Precaución: For research use only

Formula: C₇H₄BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD08457185
CAS Number: 1009094-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR