5-Fluoro-2,4-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61277-1G 61277-500MG 61277-5G 61277-25G
77472-176EA 350.2 USD
77472-176 77484-802 77495-346 77504-718
5-Fluoro-2,4-dimethoxybenzaldehyde ≥97%
5-Fluoro-2,4-dimethoxybenzaldehyde

Precaución: For research use only

Formula: C₉H₉FO₃
MW: 184.1643 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24677491
CAS Number: 155831-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR