6-Methoxy-N,N-dimethylpyridin-2-amine ≥95%

Supplier: AOBChem USA

Warning

61168-1G 61168-500MG 61168-5G 61168-25G
77472-172EA 417.15 USD
77472-172 77484-798 77495-342 77504-714
6-Methoxy-N,N-dimethylpyridin-2-amine ≥95%
6-Methoxy-N,N-dimethylpyridin-2-amine

Precaución: For research use only

Formula: C₈H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD22418419
CAS Number: 939960-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR