3-Chloro-2,4,6-trifluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

61110-1G 61110-500MG 61110-5G 61110-25G
77472-168EA 256.47 USD
77472-168 77484-794 77495-338 77504-712
3-Chloro-2,4,6-trifluorobenzoic acid ≥95%
3-Chloro-2,4,6-trifluorobenzoic acid

Precaución: For research use only

Formula: C₇H₂ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11845969
CAS Number: 174312-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR