2-Bromo-5-chloro-1-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60904-1G 60904-500MG 60904-5G 60904-25G
77472-164EA 209.09 USD
77472-164 77484-790 77495-334 77504-710
2-Bromo-5-chloro-1-iodo-3-methylbenzene ≥97%
2-Bromo-5-chloro-1-iodo-3-methylbenzene

Precaución: For research use only

Formula: C₇H₅BrClI
Storage Temperature: Ambient
MDL Number: MFCD34760019
CAS Number: 2384676-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR