Ethyl 5-(tert-butyl)-2-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

60861-1G 60861-500MG 60861-5G 60861-25G
77472-160EA 353.29 USD
77472-160 77484-786 77495-330 77504-706
Ethyl 5-(tert-butyl)-2-isopropoxybenzoate ≥97%
Ethyl 5-(tert-butyl)-2-isopropoxybenzoate

Precaución: For research use only

Formula: C₁₆H₂₄O₃
MW: 264.3599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31771668
CAS Number: 1369120-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR