1,5-Dibromo-2-iodo-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

60754-1G 60754-500MG 60754-5G 60754-25G
77472-156EA 415.09 USD
77472-156 77484-782 77495-326 77504-704
1,5-Dibromo-2-iodo-4-(trifluoromethyl)benzene ≥97%
1,5-Dibromo-2-iodo-4-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₇H₂Br₂F₃I
MW: 429.7984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759919
CAS Number: 2383685-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR