Ethyl 3-formyl-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

60622-1G 60622-500MG 60622-5G
77472-152EA 476.89 USD
77472-152 77484-778 77495-318
Ethyl 3-formyl-5-(trifluoromethyl)benzoate ≥97%
Ethyl 3-formyl-5-(trifluoromethyl)benzoate

Precaución: For research use only

Formula: C₁₁H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD22543860
CAS Number: 1018473-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR