2-(2-Bromo-5-chloro-4-methoxyphenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

60468-1G 60468-500MG 60468-5G 60468-25G
77472-148EA 197.76 USD
77472-148 77484-774 77495-314 77504-694
2-(2-Bromo-5-chloro-4-methoxyphenyl)-2-methyl-1,3-dioxolane ≥97%
2-(2-Bromo-5-chloro-4-methoxyphenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₂BrClO₃
MW: 307.568 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759761
CAS Number: 2764733-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR