1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2,2-dichloroethanol ≥95%

Supplier: AOBChem USA

Warning

60401-1G 60401-500MG 60401-5G 60401-25G
77472-144EA 415.09 USD
77472-144 77484-770 77495-308 77504-690
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2,2-dichloroethanol ≥95%
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2,2-dichloroethanol

Precaución: For research use only

Formula: C₉H₈BrCl₂FO₂
MW: 317.967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34814090
CAS Number: 2969207-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR