(3-Bromo-5-fluoro-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

60330-1G 60330-500MG 60330-5G 60330-25G
77472-140EA 284.28 USD
77472-140 77484-766 77495-302 77504-682
(3-Bromo-5-fluoro-2-methylphenyl)(methyl)sulfane ≥97%
(3-Bromo-5-fluoro-2-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₈BrFS
MW: 235.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759692
CAS Number: 2705664-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR