2-(5-Bromo-2-fluoro-3-isopropoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

60192-1G 60192-500MG 60192-5G 60192-25G
77472-136EA 428.48 USD
77472-136 77484-762 77495-298 77504-678
2-(5-Bromo-2-fluoro-3-isopropoxyphenyl)-1,3-dioxolane ≥97%
2-(5-Bromo-2-fluoro-3-isopropoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₂H₁₄BrFO₃
MW: 305.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759623
CAS Number: 2764729-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR