4,5-Dimethoxy-2-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60022-1G 60022-500MG 60022-5G
77472-132EA 653.02 USD
77472-132 77484-758 77495-294
4,5-Dimethoxy-2-(trifluoromethyl)benzaldehyde ≥97%
4,5-Dimethoxy-2-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28534102
CAS Number: 1785147-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR