4-Methyl-5-(methylthio)picolinaldehyde ≥97%

Supplier: AOBChem USA

Warning

59862-1G 59862-500MG 59862-5G
77472-124EA 697.31 USD
77472-124 77484-750 77495-286
4-Methyl-5-(methylthio)picolinaldehyde ≥97%
4-Methyl-5-(methylthio)picolinaldehyde

Precaución: For research use only

Formula: C₈H₉NOS
MW: 167.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29123824
CAS Number: 1289036-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR