1-(Benzyloxy)-2-fluoro-3-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59819-1G 59819-500MG 59819-5G 59819-25G
77472-120EA 287.37 USD
77472-120 77484-746 77495-282 77504-666
1-(Benzyloxy)-2-fluoro-3-iodo-5-methylbenzene ≥97%
1-(Benzyloxy)-2-fluoro-3-iodo-5-methylbenzene

Precaución: For research use only

Formula: C₁₄H₁₂FIO
MW: 342.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759425
CAS Number: 2807466-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR