2-(3-Bromo-2-isopropoxy-5-isopropylphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

59593-1G 59593-500MG 59593-5G
77472-116EA 438.78 USD
77472-116 77484-742 77495-278
2-(3-Bromo-2-isopropoxy-5-isopropylphenyl)-1,3-dioxolane ≥95%
2-(3-Bromo-2-isopropoxy-5-isopropylphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₅H₂₁BrO₃
MW: 329.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759258
CAS Number: 2807439-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR