2-(4-Bromo-2-ethyl-6-methylphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

59512-1G 59512-500MG 59512-5G 59512-25G
77472-112EA 415.09 USD
77472-112 77484-738 77495-274 77504-662
2-(4-Bromo-2-ethyl-6-methylphenyl)-1,3-dioxolane ≥95%
2-(4-Bromo-2-ethyl-6-methylphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₂H₁₅BrO₂
MW: 271.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759204
CAS Number: 2901098-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR