Methyl 2,5-dichloro-4-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

59481-1G 59481-500MG 59481-5G
77472-108EA 1109.31 USD
77472-108 77484-734 77495-270
Methyl 2,5-dichloro-4-isopropoxybenzoate ≥97%
Methyl 2,5-dichloro-4-isopropoxybenzoate

Precaución: For research use only

Formula: C₁₁H₁₂Cl₂O₃
MW: 263.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759187
CAS Number: 2807467-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR