3-(2,4-Difluorophenyl)-3-hydroxypropanoic acid ≥95%

Supplier: AOBChem USA

Warning

59388-1G 59388-500MG 59388-5G
77472-104EA 844.6 USD
77472-104 77484-730 77495-266
3-(2,4-Difluorophenyl)-3-hydroxypropanoic acid ≥95%
3-(2,4-Difluorophenyl)-3-hydroxypropanoic acid

Precaución: For research use only

Formula: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD16749138
CAS Number: 1248626-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR