4-Fluoro-3-(trimethylsilyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

59380-1G 59380-500MG 59380-5G 59380-25G
77472-100EA 415.09 USD
77472-100 77484-726 77495-262 77504-658
4-Fluoro-3-(trimethylsilyl)benzonitrile ≥97%
4-Fluoro-3-(trimethylsilyl)benzonitrile

Precaución: For research use only

Formula: C₁₀H₁₂FNSi
MW: 193.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759131
CAS Number: 2708175-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR