2-(2-Fluoro-5-(methylthio)phenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

59333-1G 59333-500MG 59333-5G 59333-25G
77472-096EA 163.77 USD
77472-096 77484-722 77495-258 77504-654
2-(2-Fluoro-5-(methylthio)phenyl)-1,3-dioxolane ≥95%
2-(2-Fluoro-5-(methylthio)phenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₁FO₂S
MW: 214.257 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759117
CAS Number: 2764732-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR