4-Iodo-1-(methoxymethoxy)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

59103-1G 59103-500MG 59103-5G 59103-25G
77472-088EA 229.69 USD
77472-088 77484-714 77495-250 77504-648
4-Iodo-1-(methoxymethoxy)-2-methylbenzene ≥95%
4-Iodo-1-(methoxymethoxy)-2-methylbenzene

Precaución: For research use only

Formula: C₉H₁₁IO₂
Storage Temperature: Ambient
MDL Number: MFCD34758999
CAS Number: 2807470-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR